Learn More
Fisherbrand™ Cotton-Tipped Applicators
Purified cotton
 View more versions of this product
Catalog No. 23-400-105
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
Wood
7.62cm
Cotton
Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens and for cleaning delicate instruments
3 in.
No
Swab