Learn More
Fisherbrand™ Cover Glasses: Circles
Made of finest optical borosilicate glass, with uniform thickness and size
 View more versions of this product
Catalog No. 22-293232
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Made of finest optical borosilicate glass, with uniform thickness and size
Circular shape.
  • Corrosion-resistant
  • Lint-free and ready to use
  • Available in squares, circles and rectangles
Order Info
Packed in 1 oz. (28g) plastic boxes with custom-molded bases for easy one-at-a-time dispensing
Specifications
Specifications
Borosilicate Glass
12mm
Microscopes
Circle
0.17 to 0.25mm
Coverglass