Fisherbrand™ Cover Glasses: Circles Available on GSA/VA Contract for Federal Government customers only.
Made of finest optical borosilicate glass, with uniform thickness and size
 View more versions of this product
Catalog No. 22-031145
  Check Availability
Add to Cart
    Specifications
    Product Type Coverglass
    Material Borosilicate Glass
    Shape Circle
    Diameter (Metric) 15mm
    Thickness (Metric) 0.17 to 0.25mm
    For Use With Microscopes
  • Description & Specifications
Related Products
Product Content Feedback