Fisherbrand™ Cover Glasses: Circles
Made of finest optical borosilicate glass, with uniform thickness and size
 View more versions of this product
Catalog No. 22-031144
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type Coverglass
    Material Borosilicate Glass
    Shape Circle
    Diameter (Metric) 15mm
    Thickness (Metric) 0.13 to 0.17mm
    For Use With Microscopes
  • Description & Specifications
Related Products
Product Content Feedback