Learn More
Fisherbrand™ Cover Glasses: Rectangles
Made of finest optical borosilicate glass, with uniform thickness and size
This item is not returnable.
View return policy
Description
Made of finest optical borosilicate glass, with uniform thickness and size
Rectangular shape.
- Corrosion-resistant
- Lint-free and ready to use
Order Info
Approximately 66 pieces packed in 1 oz. (28g) plastic boxes with custom-molded bases for easy one-at-a-time dispensing
Specifications
Cover Glass | |
Borosilicate Glass | |
60 mm | |
Microscope, histology, cytology, hematology, microbiology |
Rectangle | |
24 mm | |
#1.5 (0.16 to 0.19 mm) |