Learn More
Fisherbrand™ Cover Glasses: Rectangles
Made of finest optical borosilicate glass, with uniform thickness and size
Description
Made of finest optical borosilicate glass, with uniform thickness and size
Rectangular shape.
- Corrosion-resistant
- Lint-free and ready to use
Order Info
Approximately 66 pieces packed in 1 oz. (28g) plastic boxes with custom-molded bases for easy one-at-a-time dispensing
Specifications
Cover Glass | |
Borosilicate Glass | |
30 mm | |
Microscope, histology, cytology, hematology, microbiology |
Rectangle | |
24 mm | |
0.17 to 0.25 mm |