Learn More
Fisherbrand™ Cover Glasses: Rectangles Available on GSA/VA Contract for Federal Government customers only.
Made of finest optical borosilicate glass, with uniform thickness and size
 View more versions of this product
Catalog No. 22-266973
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Made of finest optical borosilicate glass, with uniform thickness and size
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Approximately 66 pieces packed in 1 oz. (28g) plastic boxes with custom-molded bases for easy one-at-a-time dispensing
Specifications
Specifications
Borosilicate Glass
24mm
0.17 to 0.25mm
Coverglass
Rectangular
60mm
Microscope, histology, cytology, hematology, microbiology