Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Cross-Type Stir Bars Available on GSA/VA Contract for Federal Government customers only.

Agitate liquids in round-bottom test tubes and flasks, where other stir bars cannot be used.

 View more versions of this product

Catalog No. 14-512-165



Add to cart

Description

Description

Agitate liquids in round-bottom test tubes and flasks, where other stir bars cannot be used.

  • Maintains its position in a vessel and will not wobble
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

50
22.2mm
Cross
ISO 9001:2008
White
Round bottom test tubes and flasks
12.7mm
Stir bar
Magnet
Yes
PTFE
0.88 in.
0.5 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit