Learn More
Fisherbrand™ Crucibles
Easy to clean smooth, glazed surface.
$45.75 - $118.00
Specifications
Material | Porcelain |
---|---|
Finish | Smooth, Glazed |
For Use With (Application) | Used in chemical environments |
Max. Temperature (Metric) | 1150°C |
Catalog Number | Capacity (Metric) | Product Type Specs | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Capacity (Metric) | Product Type Specs | Price | Quantity & Availability | |||||
S02224
|
50 mL | High-form |
Pack of 6 for $96.00
Case of 8 PK for $694.00
|
|
|||||
S02221
|
15 mL | High-form |
Pack of 6 for $49.50
Case of 8 PK for $359.00
|
|
|||||
S02222
|
30 mL | High-form |
Pack of 6 for $74.25
Case of 8 PK for $540.00
|
|
|||||
S02220
|
10 mL | High-form |
Pack of 6 for $45.75
Case of 8 PK for $332.00
|
|
|||||
S02225
|
100 mL | High-form |
Pack of 6 for $103.00
Case of 8 PK for $745.00
|
|
|||||
S02227
|
17 mL | Wide-form |
Pack of 6 for $53.00
Case of 8 PK for $385.00
|
|
|||||
S02230
|
100 mL | Wide-form |
Pack of 6 for $89.00
Case of 8 PK for $647.00
|
|
|||||
S02228
|
30 mL | Wide-form |
Pack of 6 for $61.50
Case of 8 PK for $448.00
|
|
|||||
S02231
|
250 mL | Wide-form |
Pack of 6 for $118.00
Case of 8 PK for $852.00
|
|
|||||
Description
- Use in any chemical environment
- Withstand temperatures up to 1150°C (2,102°F)
- Consistent wall thickness and density
- Fired twice to give added strength and durability
- Highly resistant to chemical attack
Order Info
Crucible covers sold separately (Cat. Nos. S02232 through -238).
Specifications
Porcelain | |
Smooth, Glazed | |
Used in chemical environments | |
1150°C |