Learn More
Fisherbrand? Cryo/Freezer Boxes - PROMO
Low-cost storage for laboratories in universities, government, medical research, and pharmaceutical facilities
 View more versions of this product
Catalog No. 03-395-465PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
5 x 5 x 2 in. (12.7 x 12.7 x 5.1cm)
Provides low-cost storage for laboratories in universities, government, medical research, and pharmaceutical facilities; in institutions performing artificial insemination, biotech, blood bank, and stem cell/bone marrow procedures
Fiberboard
100
-320.8°F
White
Used in mechanical freezers and liquid nitrogen tanks and all standard stainless steel and polycarbonate storage systems
Bottom drain holes, tight lid
-196°C