Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Cryovial™ Storage Boxes
Durable, stackable polycarbonate storage boxes for vials from 1 to 5mL
Description
- Withstand temperatures from -196° to +121°C (-321° to +250°F)
- On 25- and 81-place boxes, a colored plastic grid insert separates the cover from the base
- The 100-place boxes feature a colored base instead of a grid insert
- Transparent cover allows view of box contents and is keyed to the base to prevent misalignment
- All boxes are supplied with a vial-removal tool to facilitate removal of vials from a full box
- Autoclavable
Specifications
Yellow | |
81 tubes | |
4/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction