Learn More
Fisherbrand™ Deuterium Lamps for Background Correction
Alert:
Warning:for safe operation of deuterium lamps, the operator should never look directly at the lamp. Products produce high levels of deep UV light, and it is imperative that appropriate eye protection is used. Also, the envelope can reach 200°C during operation, and proper precautions must be taken if lamp is to be handled.
Products 2
Catalog Number Description Price Qty Availability  
Catalog Number Description Price Qty Availability  
14-386-25L
View Documents
N/A 28 to 30mm OD This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-386-25R
View Documents
N/A 28 to 30mm OD This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A