Learn More
Fisherbrand™ Diamond Tip Pencils Non-distribution product as customer accommodation. Get IT Item
For microscopic identification and on X-ray film, glass, plastics, vitreous surfaces, ceramic tiles and metals
Manufacturer:  Fisherbrand™ PENCIL DIAMD
 View more versions of this product
Catalog No. 22-268912
Add to cart
Description
Description
  • Ground polished 90° point
  • Aluminum
  • Point fused to steel stylus
  • Sharp/clean lines
Specifications
Specifications
Diamond tip pencil
Used on glass, plastics, vitreous surfaces, ceramic tiles, metals, microscopic identification and on X-ray film
Aluminum