Learn More
Fisherbrand™ DISPENSE Filter System Available on GSA/VA Contract for Federal Government customers only.
Saves time, reduces lab costs and increases safety.
 View more versions of this product
Catalog No. 22-288431
Add to cart
Description
Description
The restricted opening provides effective specimen closure and can help minimize exposure to droplets of blood or other potentially infectious materials during sample handling, preparation, processing, and storage.
  • All-inclusive “closed” filtration system with a combination filter, transfer pipet, and cap
  • Filters fibrin and gel particulates, dispenses serum or plasma, prevents evaporation, and helps protect ISE systems
  • All-in-one system maintains sample integrity and eliminates transfer error
  • Dispense minute or large amounts of sample without transfer pipets or add-on products
  • Ideal for dispensing samples for instrumentation, pour-off or other use
  • Use with glass collection tubes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
23
Use with Glass collection tubes
100 Pack
Dispensing samples for instrumentation, pour-off or other use
Combination filter, transfer pipet, cap
Dispense Filtration System