Learn More
Fisherbrand™ Dispenser Tips for Handystep™ S Repeating Pipetter
Manufactured from high-quality virgin plastics for use with most standard repeating pipettes in a wide variety of applications. The tips are made without chemical additives to prevent reagent contamination.
Description
Manufactured from high-quality virgin plastics for use with all standard repeating pipettes in a wide variety of applications. The tips are made without chemical additives to prevent reagent contamination.
25 mL and 50 mL tips require the use of an adapter. One reusable adapter supplied in each package of 25 mL and 50 mL tips.
- Precision seal between the piston and cylinder ensure smooth operation, increased accuracy and precision
- Extended piston design minimizes dead volume and maximizes fluid utilization
- Made entirely from high-quality virgin, high density polyethylene and polypropylene (piston of 0.1 mL tip is liquid crystal polymer-LCP)
Available sterile or nonsterileFits these models but not limited to:
- BRAND™ HandyStep™, HandyStep™ S, HandyStep™ electronic, HandyStep™ touch, HandyStep™ touch S
- All previous Fisherbrand Repeating Pipettes
- Rainin AutoRep™ M, AutoRep™ S, AutoRep™ E
- Eppendorf Multipette™ 4780
- Gilson Repetman™
- DLAB StepMate
- Scilogex Repeater
Specifications
50 mL | |
Non-Filtered | |
Translucent | |
≤0.4 to 0.15% | |
Virgin raw materials | |
Positive Displacement | |
Single-use | |
Yes | |
Printed |
10000 to 50000μL | |
Non-sterile | |
Not Autoclavable | |
±≤1.5 to 0.3% | |
Bagged | |
25/Pk. | |
Most repeating pipette models (see Description) | |
HDPE, Polypropylene |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.