Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Disposable Cuvettes

Choice of polystyrene or methacrylate

 View more versions of this product

Catalog No. 14-955-130

  Check Availability
Add to cart

Specifications

Description

Fisherbrand™ Disposable Cuvettes are available in choice of polystyrene or methacrylate. Select polystyrene cuvets for assays in visible spectral range (340 to 750nm); for accuracy throughout UV/Vis range (285 to 750nm), choose cuvets made from a superior grade of methacrylate.

  • Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
  • Choice of 4.5 and 1.5mL sample capacity
  • Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
  • Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
  • Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity
  • Each case contains cuvets with same mold impression number—assuring precision between samples—standard deviation is <±0.005 extinction units between cuvets

Description & Specifications

Specifications

45mm
Four-clear-sided
10mm
12mm
500 Case
12mm
4.5mL
Yes
For assays in visible spectral range, accuracy throughout UV/Visible range
Polymethyl methacrylate
285 to 750nm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit