Learn More
Fisherbrand™ Disposable Filter Concentrators
Ensure consistent high-quality slide preps
Description
Save time and improve results by eliminating the need to clean and sterilize reusable sample chambers
- Cells from body fluids are concentrated onto microscope slides with a level of cell recovery, clarity, and detail that cannot be achieved with direct smear techniques
- Unique integral filter card absorbs fluid at a controlled rate during spin cycle
- Snap seal for simple operation
- Hinged backing plate simplifies assembly
- Ease of use allows rapid turnaround time
Use in combination with StatSpin™ CytoFuge™ 2 and CytoFuge 12 Centrifuges
Specifications
StatSpin CytoFuge 2; CytoFuge 12 Centrifuges | |
48/Pk. |