Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm)

Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Fisherbrand? Disposable Inoculating Loops and Needles, Length: 8 in. (20cm) are precision molded from a specially formulated, biologically inert polymer.

 View more versions of this product

Catalog No. 22-363-602



 
Add to cart

Specifications

Description

Ideal for anaerobic microbiology applications where deep agar inoculation is required. Use in situations where flaming is not practical, such as under hoods and in anaerobic chambers. Flexible enough to facilitate streaking, yet rigid enough to penetrate the media for subsurface inoculation.
 
  • Combi Loop™ has a 10µL loop on one end and a 1µL loop on the other
  • Convenient than metal loops; use a loop directly from the package, then throw it away
  • Gamma irradiated for sterility in accordance with U.S. Pharmacopeia Volume XX
  • No flaming required, eliminating the hazards of open flames, spattering, cross-contamination, and the risk of infection from pathogens distributed by aerosol formation
  • Color coded for easy identification
Description & Specifications

Specifications

1/10μL
Natural
500 Pack per Case
0.09/0.38cm
Certificate of dimensional accuracy
20cm
5000/Cs.
Yes
Flexible Inoculating Combiloop
10
0.035/0.14 in.
Yes
8 in.
Biologically Inert Polymer
Ziplock Bag
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit