Learn More
Fisherbrand™ Disposable Inoculating Loops and Needles
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm) are precision molded from a specially formulated, biologically inert polymer.
$76.65 - $228.65
Specifications
Length (English) | 8 in. |
---|---|
Length (Metric) | 20 cm |
Material | Biologically Inert Polymer |
Sterility | Sterile |
Disposable | Yes |
Includes: Certificate of dimensional accuracy
Catalog Number | Product Type | Volume | Packaging Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Product Type | Volume | Packaging Type | Quantity | Price | Quantity & Availability | |||
22-363-602
|
Inoculating Loop and Needle, Flexible | 0.1 μL | Ziplock Bag | 5000/Cs. |
Pack of 500
for $76.65
Save 26% Reg :
Case of 10 PK
for $660.65
Save 26% Reg : |
|
|||
22-363-605
|
Inoculating Loop and Needle, Rigid | 10 μL | Individually wrapped | 1 Ea. |
Case of 250
for $149.65
Save 17% Reg : |
|
|||
22-363-604
|
Inoculating Loop and Needle, Rigid | 1 μL | Ziplock Bag | 9600/Cs. |
Pack of 960
for $179.65
Save 25% Reg :
Case of 10 PK
for $1,616.65
Save 25% Reg : |
|
|||
22-363-606
|
Inoculating Loop and Needle, Rigid | 10 μL | Ziplock Bag | 40 Pk. per Cs. |
Case of 1000
for $228.65
Save 19% Reg : |
|
|||
22-363-607
|
Inoculating Loop and Needle, Rigid | 10 μL | Ziplock Bag | 9600/Cs. |
Pack of 960
for $199.65
Save 20% Reg :
Case of 10 PK
for $1,797.65
Save 20% Reg : |
|
|||
22-363-603
|
Inoculating Loop and Needle, Rigid | 1 μL | Paper Peel Pouch, Individually Wrapped | 250/Cs. |
Case of 250
for $98.65
Save 23% Reg : |
|
|||
Description
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm) are precision molded from a specially formulated, biologically inert polymer.
Ideal for anaerobic microbiology applications where deep agar inoculation is required. Use in situations where flaming is not practical, such as under hoods and in anaerobic chambers. Flexible enough to facilitate streaking, yet rigid enough to penetrate the media for subsurface inoculation.- Combi Loop™ has a 10μL loop on one end and a 1μL loop on the other
- Convenient than metal loops; use a loop directly from the package, then throw it away
- Gamma irradiated for sterility in accordance with U.S. Pharmacopeia Volume XX
- No flaming required, eliminating the hazards of open flames, spattering, cross-contamination, and the risk of infection from pathogens distributed by aerosol formation
- Color coded for easy identification
Specifications
8 in. | |
Biologically Inert Polymer | |
Yes |
20 cm | |
Sterile | |
Certificate of dimensional accuracy |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.