Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Fisherbrand™ Disposable Pestle System
Low-cost alternative to mechanical or metal and glass tissue grinders
 View more versions of this product
Catalog No. 03-392-103
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Pestle Grinder System (03-392-106) comes with pestle/mixer adapter and two “AA” batteries. Disposable pestles and tubes sold separately. System components also available separately.
Description
Description
Lightweight, cordless mixer can be used with disposable pestles and microtubes
  • High-speed vortexing action completely homogenizes cells and tissue within seconds; re-suspends protein and DNA pellets
  • Battery-powered unit greatly reduces hand fatigue
  • Grooved handle makes it easy to turn, even when wearing gloves
  • Rugged polybutylene terephthalate (PBTP) plastic pestle yields superior homogenization
  • Disposable pestles and tubes can be used by themselves or in conjunction with the grinder
  • Pestles and tubes are individually wrapped to prevent cross-contamination of samples
  • DNase, RNase, and pyrogen-free