Fisherbrand™ Disposable Plain-End Glass Tubes with Color-Coded Label Area Available on GSA/VA Contract for Federal Government customers only.

Eliminate the problem of lost or peeling labels

 View more versions of this product

Catalog No. 14-957-12E

  Check Availability
Add to Cart

    Specifications

    Color Blue
    Disposable Yes
    For Use With (Application) End problem of lost or peeling labels
    Diameter (Metric) Outer 12mm
    Length (Metric) 75mm
    Material Flint Glass
    Shape Round

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit