Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Disposable Plastic Cuvets

Choice of polystyrene or methacrylate

 View more versions of this product

Catalog No. 14-385-985

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
  • Select polystyrene cuvets for assays in visible spectral range (340 to 750nm)
  • For measurements in the visible and near-UV range (300-750nm) choose cuvets made from polymethyl methacrylate (acrylic)
  • Choice of 4.5 and 1.5mL sample capacity
  • Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
  • Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
  • Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity

Order Info

  • Packaged 100 cuvets per stackable tray; five trays to a case
  • Outside dimensions: 12L x 12W x 45mmH

Specifications

Specifications

Polystyrene
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit