Fisherbrand™ Disposable Polypropylene Spatulas

Economical, single-use lab tools that scoop, scrape and stir

 View more versions of this product

Catalog No. 14-955-551

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »

Disclaimers:

Fisher #14-955-550, 14-955-551 and 14-955-553 are licensed under U.S. Patent 8,709,362


    Specifications

    Autoclavable Yes
    Color Opaque
    Disposable Yes
    For Use With (Application) Ideal for transferring, collecting, and handling chemical and biological materials
    Includes Double end with spatula/spoon on one end and a smaller scoop on other end, hallow core
    Length (English) 8.26 in.
    Length (Metric) 21cm
    Material Polypropylene
    Size Standard
    Product Type Spatula

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit