Learn More
Fisherbrand Disposable Soda Lime Glass Pipets
Ideal for transferring samples to sample cups and for drop counting.
Description
- Carton top folds back tilts carton into convenient dispensing position
- Tip openings range from 0.80 to 1.20mm
- Top and tip free of sharp edges
- Can be cotton plugged (plugs not included)
Specifications
Yes | |
Yes | |
5.75 in. | |
Soda Lime Glass | |
2 mL |
0.80 to 1.20 mm | |
Uniform bore, free or sharp edge (tip and top) | |
14.6 cm | |
Pasteur Pipet |