Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Durac™ Plus™ Plastic Case Pocket Liquid-In-Glass Thermometers

With accuracy of 1.5°C or better

 View more versions of this product

Catalog No. 13-201-683


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.5°C
Suitable for field studies, waste water treatment and any portable applications
Window plastic case
160mm
0.5°C
-5° to +50°C
ISO/IEC 17025:2005, A2LA, NIST
Total
6.3 in.
Glass
Not Applicable
Blue Organic
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit