Fisherbrand™ Dye Industry Pipets Available on GSA/VA Contract for Federal Government customers only.
Perfect for pipetting dark-color dye solutions
 View more versions of this product
Catalog No. 13-671-109H
  Check Availability
Add to Cart
    Specifications
    Accuracy ±0.02mL
    Calibration To deliver
    Color Code Green
    Color Green
    Disposable No
    Graduated Yes
    Packaging Quantity Case of 12
    Subdivision 0.1mL
    Capacity (Metric) 2mL
    Material Borosilicate Glass
    Product Type Dye Industry Pipet
  • Description & Specifications
Related Products
Product Content Feedback