Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Economy Grade Latex Coated Palm Gloves
Click to view available options
Size:
Small
Large
Description
- Red latex, smooth coated palm
- Coated palm offers abrasion resistance and increased grip
- Uncoated back allows hands to breathe
- Ideal for assembly, construction, inspection, material handling, refuse handling, and landscaping
Specifications
Coated Area | Palm |
Coating Material | Latex |
Color | Natural |
Grade | Economy |
Material | Polycotton |
Product Type | Dip-coated Gloves |
Size | Large |
For Use With (Application) | Assembly, Construction, Inspection, Material Handling, and Landscaping |
Glove Cuff Style | Continuous Knit |
Series | 39-C121 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction