Fisherbrand™ Electronic Pipetters
Electronic tip ejection combined with unique ergonomic design
 View more versions of this product
Catalog No. 21-165-87
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Part Not Found  Please contact customer service at 1-800-766-7000 with error code:999. 
Required Accessories : Must order carousel charging stand (sold separately).
    Specifications
    Accuracy 4 to 0.9%
    Battery Type NiMH
    Compatible Tips Universal fit pipette tip
    Number of Channels 12
    Display LCD
    Volume (Metric) Range 0.2 to 10μL
    Volume (Metric) 0.2 to 10μL
    Finger Style Ergonomic
    For Use With (Equipment) Universal fit pipette tip
    Increments 0.05μL
    User-friendly design takes the effort out of pipetting and reduces the risk of work-related upper limb disorders that are common in mechanical pipetting. Choose single, 8-, and 12-channel models in a variety of volume ranges.
    • Clear LCD readout and easy-to-use keypad
    • Electronic tip-ejection buttons, located for left- or right-handed operation, eliminate tip release pressure
    • User-programmable modes, storable routine pipetting protocols, and fully adjustable speed settings for all operations
    • Electronic start button activates all piston actions, reducing strain on thumb joints and carpal tunnel Computerized drive system lets you work more efficiently than with mechanical or other electronic pipetters Replaceable tip cone filter on most models helps minimize risk of internal contamination
    • Operations include pipetting, reverse pipetting, pipetting with mixing, pipetting with cycle counter, manual pipetting, multi-dispensing, diluting, diluting with mixing, sequential dispensing, multi-aspirating, and automatic multi-dispensing.
    • Can be charged using single-pipetter charging stand, 4-pipetter carousel charging stand, or AC-charging adapter; replacement NiMH battery packs are available (FS Cat. No. 21-390-45)
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback