Fisherbrand™ Electronic Pipetters
Electronic tip ejection combined with unique ergonomic design
 View more versions of this product
Catalog No. 21-165-88
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Required Accessories : Must order carousel charging stand (sold separately).
    Specifications
    Accuracy 2 to 0.5%
    Battery Type NiMH
    Compatible Tips Universal fit pipette tip
    Display LCD
    Finger Style Ergonomic
    For Use With (Equipment) Universal fit pipette tip
    Increments 0.5μL
    Number of Channels 12,12
    Volume (Metric) Range 5 to 120μL
    Volume (Metric) 5 to 120μL
    Product Type Electronic Pipetter
    Capacity Range 5 to 120μL
    No. of Positions 12.0
    Compatibility Universal fit pipette tip
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback