Fisherbrand™ Electronic Pipetters
Electronic tip ejection combined with unique ergonomic design
 View more versions of this product
Catalog No. 21-165-82
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Required Accessories : Must order carousel charging stand (sold separately).
    Specifications
    Accuracy 1 to 0.5%
    Battery Type NiMH
    Compatible Tips Universal fit pipette tip
    Display LCD
    Finger Style Ergonomic
    For Use With (Equipment) Universal fit pipette tip
    Increments 10μL
    Number of Channels 1.0,1
    Volume (Metric) Range 100 to 5000μL
    Volume (Metric) 100 to 5000μL
    Product Type Micro
    Capacity Range 100 to 5000μL
    No. of Positions 1.0
    Compatibility Universal fit pipette tip
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback