Fisherbrand™ Electroporation Cuvettes Plus™ Promo
Disposable, color-coded cuvets fit most electroporators
 View more versions of this product
Catalog No. 98-549-0PM
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Electroporators
    Product Type Electroporation Cuvette
    Volume (Metric) 90μL
    Includes Imbedded aluminum electrodes; Case of 50
    Material Polycarbonate
    Sterile Yes
    Width (Metric) Gap 1mm
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback