Fisherbrand™ Electroporation Cuvettes Plus™ Promo
Disposable, color-coded cuvets fit most electroporators
 View more versions of this product
Catalog No. 98-549-1PM
  Check Availability
Add to Cart
    Specifications
    Material Polycarbonate
    Product Type Electroporation Cuvette
    Volume (Metric) 400μL
    Width (Metric) Gap 2mm
    For Use With (Equipment) Electroporators
    Includes Imbedded aluminum electrodes; Case of 50
    Sterile Yes
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback