Learn More
Torrey Pines Scientific™ EchoTherm™ Benchtop Chilling/Heating Incubator Sample Blocks
$314.21 - $521.49
Specifications
Material Aluminum
Product Type Block
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
11-680-31 Torrey Pines Scientific™
6205020
Echotherm benchtop chilling/heating incubator Each for $456.04
Check Availability
Add to cart
11-680-28 Torrey Pines Scientific™
6205016
Echotherm benchtop chilling/heating incubator Each for $521.49
Check Availability
Add to cart
11-680-27 Torrey Pines Scientific™
6205015
Echotherm benchtop chilling/heating incubator Each for $471.12
Check Availability
Add to cart
11-680-26 Torrey Pines Scientific™
6205014
Echotherm benchtop chilling/heating incubator Each for $433.29
Check Availability
Add to cart
11-680-25 Torrey Pines Scientific™
6205013
Echotherm benchtop chilling/heating incubator Each for $349.08
Check Availability
Add to cart
11-680-24 Torrey Pines Scientific™
6205012
Echotherm benchtop chilling/heating incubator Each for $314.21
Check Availability
Add to cart
Specifications
Description
Blocks are machined from solid aluminum to fit precisely the tube size and shape. Block bottoms are also machined to nest on the incubator plate to prevent movement. Feature an anodized black finish for better heat transfer.
Description & Specifications
Specifications
Aluminum
Block