Learn More
Fisherbrand™ Excel™ Ear Plugs
NRR: 30dB
 View more versions of this product
Catalog No. 19-070476
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Preshaped and tapered for easy insertion
  • Smooth, soil-resistant surface won't irritate the ear canal
  • Plugs roll to extremely small size and expand slowly for comfortable fit
  • Integrated flexing action adjusts to head and jaw movements—helps keep employees comfortable all day, without irritating pressure
  • One size fits all—including those with small, hard-to-fit ear canals
Compliance
Attenuation tested in accordance with ANSI™ S3.19-1974.
Specifications
Specifications
One size fits to all
Excel™ Earplug
Uncorded
ANSI S3.19-1974
Smooth, soil resistant surface won't irritate the ear canal
Tapered
30dB