Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Extended Cuff Nitrile Exam Gloves Get IT Item

Choose a glove with exceptional durability and added protection. The Fisherbrand™ Extended Cuff Nitrile Exam Gloves are extra-thick, fully textured for a superior grip and are tested for use with chemotherapy drug exposure.

 View more versions of this product

Catalog No. 19-041-170A



  Check Availability
Add to cart

Specifications

Description

Extra-thick 6mil formulation provides exceptional durability and added protection.

  • Powder-free
  • Fully textured finish for a superior wet or dry grip
  • Ambidextrous and nonsterile with beaded cuff
  • Tested for use with chemotherapy drug exposure
  • Length: 12 in. (30.5cm)
  • Thickness: 6mil (palm), 7mil (finger), 4.5mil (cuff)
  • Teal

Description & Specifications

Specifications

Textured
Ambidextrous
Small
ISO9001 QMS, NFPA, AQL 1.5, ASTM F 1671, ASTM F 739
Teal
12 in.
Long, beaded
Nitrile
6mil
Powder-free
Non-sterile
30.5cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit