Learn More
Fisherbrand™ Extended-Length Tips
Extended length tips are ideal for difficult-to-reach samples
 View more versions of this product
Catalog No. 02-681-423
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Easily reach the bottom of long tubes
  • Minimize pipet contamination by preventing pipet shaft from contacting the tube or vessel
  • Natural color
  • Certified by independent laboratory– Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
Specifications
Specifications
Lift-off Rack
100 to 1000μL
White
9.14cm
Universal Fit, Extended Length
Sterile
3.6 in.