Fisherbrand™ Facemasks
Reduce particle generation
 View more versions of this product
Catalog No. 18-960A
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Filter Efficiency 99%
    Color Blue
    Construction Thin middle layer of Meltblown Polypropylene filter media lies between Spunbond Polypropylene inner and outer layers ultrasonically welded to reduce particle generation
    Includes Earloops and clips
    Material Meltblown Polypropylene
  • Description & Specifications
Related Products
Product Content Feedback