Fisherbrand™ Facemasks
Reduce particle generation
 View more versions of this product
Catalog No. 18-960C
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Filter Efficiency 99%
    Color Blue
    Construction Thin middle layer of Meltblown Polypropylene filter media lies between Spunbond Polypropylene inner and outer layers ultrasonically welded to reduce particle generation
    Includes Ties
    Material Meltblown Polypropylene
  • Description & Specifications
Related Products
Product Content Feedback