Learn More
Fisherbrand Faceshield
Comes apart for easy disinfection.
Alert:
Faceshields should be used in conjunction with protective spectacles or goggles to ensure appropriate eye protection against chemical splash or impact.
Description
- Large, 8.5 in.H (21.6cm) molded window
- 99.9% UV protection up to 380nm
- Ledge on the headgear prevents fluid from splashing down inside the window
- Pin-lock headgear can be sterilized and disinfected; window can be steam autoclaved
Compliance
Meets ANSI Z87.1-1989 standards.
Specifications
Meets ANSI Z87.1-1989 | |
8.5 in. | |
21.6 cm |