Oak Ridge False Bottom Analyzer Tubes

Tubes are sealed with a screw-on cap

Manufacturer: OAK RIDGE PRODUCTS 02160100

 View more versions of this product

Catalog No. 22-730-431

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Suitable for direct sample analysis and are ideal for any transport and storage application
    Capacity (Metric) 5mL
    Color Natural
    Material Polypropylene
    Shape Circular, Conical Bottom
    Closure Type Screw

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit