Learn More
Fisherbrand™ Filter Adapters
SDP
Eliminates boring of special size holes in rubber stoppers
 View more versions of this product
Catalog No. 05-888-102
Edge
Add to Cart
Description
Description
Eliminates boring of special size holes in rubber stoppers
Designed to fit Buchner and fritted-glass funnels, porcelain filters without headpieces, and reverse flow cleaning of filter crucibles. Useful in filtration assemblies for supporting funnels, Gooch crucibles, and filter tubes.
  • Neoprene rubber
  • Autoclavable
  • Adapter does not need to be fitted closely around the funnel stem to be functional
  • Can be used singly or nesting with adjacent sizes
  • Available as an adapter set, or in individual sizes
Specifications
Specifications
Autoclavable
Neoprene Rubber
0.1 in.
Filter Adapter
2.5 cm
Size 3
Buchner and fritted Glass funnels, porcelain filters without headpieces
0.3 cm
0.9 in.
37 x 22 mm
Product Suggestions
Product Suggestions