Fisherbrand™ Filter Adapters
Eliminates boring of special size holes in rubber stoppers
 View more versions of this product
Catalog No. 05-888-107
  Check Availability
Add to Cart
    Specifications
    Autoclavable Yes
    Product Type Filter Adapter
    For Use With (Equipment) Buchner and fritted Glass funnels, porcelain filters without headpieces
    Material Neoprene Rubber
  • Description & Specifications
Related Products
Product Content Feedback