Fisherbrand™ Finnpipette™ II Adjustable-Volume Pipetters
Fully autoclavable for absolute sterility
 View more versions of this product
Catalog No. 21-377-821
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Secure Tip Attachment Soft touch tip ejection
    Warranty 3 Years
    Color Blue,Blue
    Product Type Micro
    Accuracy ±1 to 0.6%
    Autoclavable Yes
    Finger Style Ergonomic
    Includes Calibration certificate, in-lab calibration tool
    Increments 5μL
    Volume (Metric) Range 100 to 1000μL
    Volume (Metric) 100 to 1000μL
    Capacity Range 100 to 1000μL
    Type Micro
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback