Fisherbrand™ Finnpipette™ II Digital Multichannel Pipetters
Advanced pipetting performance in a lightweight, ergonomic design
 View more versions of this product
Catalog No. 21-377-830
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Autoclavable Yes
    Increments 5μL
    Warranty 3 Years
    Color Yellow,Yellow
    Number of Channels 12
    Volume (Metric) Range 50 to 300μL
    Volume (Metric) 50 to 300μL
    Product Type Digital Multichannel Pipetter
    Capacity Range 50 to 300μL
  • Description & Specifications
Related Products
Product Content Feedback