Fisherbrand™ Finnpipette™ II Fixed-Volume Pipetters
Superior comfort, accuracy, and precision
 View more versions of this product
Catalog No. 14-487-70
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Accuracy ±0.6%
    Autoclavable Yes
    Compatible Tips Universal fit pipette tip
    Finger Style Ergonomic
    For Use With (Equipment) Universal fit pipette tip
    Secure Tip Attachment Soft touch tip ejection
    Warranty 3 Years
    Volume (Metric) 25μL
    Product Type Fixed Volume Pipetter
    Capacity Range 25μL
    Compatibility Universal fit pipette tip
  • Description & Specifications
Related Products
Product Content Feedback