Fisherbrand™ Finnpipette™ II Fixed-Volume Pipetters
Superior comfort, accuracy, and precision
 View all options for this product
Catalog No. 14-487-73
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Accuracy ±0.4%
    Compatible Tips Universal fit pipette tip
    Autoclavable Yes
    Volume (Metric) 200μL
    Finger Style Ergonomic
    For Use With (Equipment) Universal fit pipette tip
    Secure Tip Attachment Soft touch tip ejection
    Warranty 3 Years
    Today's laboratories require sophisticated and reliable liquid handling technology that allows its users to cope with the ever-increasing demands in their daily work. This fixed-volume pipetter combines a sleek, ergonomic design, advanced features, and uncompromising performance to better respond to those needs.
    • Ideal for a variety of general pipetting applications
    • Comfortable, lightweight handle with finger rest Fully autoclavable Soft-touch tip ejector (50% reduced force) Easy in-lab calibration and service
  • Description & Specifications
Related Products
Product Content Feedback