Learn More
Fisherbrand™ Floating Test Tube Racks
Eliminate expensive plastic floating racks with economical polyfoam floating racks.
$47.50
Specifications
Product Type | Tube Rack |
---|---|
Autoclavable | Not Autoclavable |
Color | Gray |
Material | Polymer Foam |
Sterility | Non-sterile |
Description
- Economical and nonabsorbent
- Easy to use in water baths or beakers
- Inscriptions with tube marker pens may be made for quick identification
Specifications
Tube Rack | |
Gray | |
Non-sterile |
Not Autoclavable | |
Polymer Foam |