Floating Test Tube Racks

Eliminate expensive plastic floating racks with economical polyfoam floating racks

 View more versions of this product

Catalog No. 14-127-44

  Check Availability
Add to Cart

    Specifications

    Size 83 x 9.5mm (3.25 x .125 in.)
    For Use With (Equipment) 0.5 and 1.5mL microtubes,0.5 and 1.5mL Microtubes
    Autoclavable No
    Holds 0.5 and 1.5mL microtubes
    Material Polyfoam
    Type Tube Rack
    Shape Circle
    Product Type Tube Rack
    Tube Size 0.5 and 1.5mL microtubes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit