Fisherbrand™ Flowmeter Regulators [GenStar]
For Carbon Dioxide, Nitrogen and Oxygen
 View more versions of this product
Catalog No. 10-575-100
  Check Availability
Add to Cart
    Specifications
    Case Material Brass
    Connections 0.62 in.-18RH(F) and 0.56 in.-18RH(M)
    Airflow Rate 10-60 SCFH
    Pressure Rating 25psig
    For Use With (Application) Carbon Dioxide
    Temperature Rating 0° to 140°C
    Internal Part Material Neoprene Fabric Diaphragm
    Seal Material Nylon
  • Description & Specifications
Related Products
Product Content Feedback