Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Fluorescence Rectangular Cells: Micro Cells With PTFE Lids, 3 Clear Windows

Fit standard spectrophotometer cell holders

 View more versions of this product

Catalog No. 14-958-104



  Check Availability
Add to cart

Specifications

Description

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Reduce reagent use with micro cells
  • Wavelength range for all cells is 200 to 2500nm
  • Quartz windows for transmission from far ultraviolet through the infrared

Description & Specifications

Specifications

45μL
For transmission from far ultraviolet through the infrared
45mm
Ea., 4 Ea./Cs.
Rectangular
12.5mm
Quartz
PTFE
Standard spectrophotometer cell holder
8.5mm
3mm
200 to 2500nm
3mm
Micro Fluorescence Cell
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit